Order Entry
Denmark
Orders LinkContactUsLinkComponent
L(+)-Glutamic acid, Millipore®

L(+)-Glutamic acid, Millipore®

L(+)-Glutamic acid, Millipore®
Supplier:  Merck
Certificates
L(+)-Glutamic acid, Millipore®
L(+)-Glutamic acid, Millipore®

3 Options Available Below

Product Details & Documents

Soluble in water and mineral acids; insoluble in organic solvents. In aqueous solutions forms pyrrolidonecarboxylic acid slowly at room temperature, more rapidly at 100 °C.

Synonyms:  (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
  • CAS Number:56-86-0
  • Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00002634
  • Molecular Weight:147.13 g/mol
  • Merck Index:13,04482
  • Boiling Pt:325 °C (1013 hPa)
  • Melting Pt:160 °C
  • Density:1.538 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-293-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 724,00 kr
    724,00 kr
    1 * 250 g
       724,00 kr

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 2.420,00 kr
    2.420,00 kr
    1 * 1 kg
       2.420,00 kr

  • Plastic drum
    10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 18.420,00 kr
    18.420,00 kr
    1 * 10 kg
       18.420,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.