Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

L(+)-Glutamic acid

Supplier:   PanReac AppliChem
L(+)-Glutamic acid
Supplier:  PanReac AppliChem
Certificates
L(+)-Glutamic acid
L(+)-Glutamic acid

2 Options Available Below

Product Details & Documents
Synonyms:  (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
  • CAS Number:56-86-0
  • Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00002634
  • Molecular Weight:147.13 g/mol
  • Merck Index:13,04482
  • Boiling Pt:325 °C (1013 hPa)
  • Melting Pt:160 °C
  • Density:1.538 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-293-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1.500,00 kr
    1.500,00 kr
    1 * 1 kg
       1.500,00 kr

  • 10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg