Sodium gluconate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: D-Gluconic acid sodium salt, Sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic acid, sodium salt, Sodium D-gluconate
CAS Number:527-07-1
:
Formula:HOH₂C(CH(OH))₄COONa
MDL Number:MFCD00064210
Molecular Weight:218.14 g/mol
Melting Pt:∼ 206 °C (dec.)
Merck Index:12,08766
Storage Temperature:Ambient
EINECS:208-407-7
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together