Powder spatulas, disposable, SteriWare®
PP. These disposable spatulas feature a rounded handle and a robust blade.
- Manufactured and packed in a controlled atmosphere (class 100000 cleanroom)
- Cross-contamination risk is eliminated - the device can be used straight from the bag
- Approved for use in ATEX Zones 0, 1 & 2 (contact Sampling Systems for full details)
- Disposable: No cleaning is required before or after sampling
- Available as gamma irradiated sterile (SAL 10⁻⁶)
Suitable for sterile sampling and dispensing quantities of powders in all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.).
Frequently Bought Together