Order Entry
Denmark
ContactUsLinkComponent
 
D-(-)-Diethyl tartrate for synthesis, Sigma-Aldrich®
  8.18685.0025
 :  Merck
 :  
D-(-)-Diethyl tartrate for synthesis, Sigma-Aldrich®
  8.18685.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    17 °C
  • Molar Mass:
    206.19 g/mol
  • Boiling Point:
    162 °C (25 hPa)
  • Vapor Pressure:
    93 °C
  • Flash Point:
    1.4467 (20 °C, 589 nm)
  • Density:
    1.20 g/cm3 (20 °C)
  • Synonyms:
    L-(+)-Tartaric acid diethyl ester, DET, (-)-Diethyl-D-tartrate
  • Hill Formula:
    C8H14O6
  • Chemical Formula:
    C2H5OOC(CHOH)2COOC2H5

 

 

 :
 :280 °C (1013 hPa)
 :17 °C
 :1.2 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :206.2 g/mol
 :237-458-8
 :13811-71-7
 :C₈H₁₄O₆
 :MFCD00064451