Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Amphotericin B

250 mg
Catalog # P4030-250MG
Supplier:   Biowest
Amphotericin B
Catalog # P4030-250MG
Supplier:  Biowest
CAS Number:  1397-89-3
Amphotericin B
Amphotericin B

250 mg

CAS Number:  1397-89-3
Price
470,00 kr
Log in for availability
1 * 250 mg
Product Details & Documents

Amphotericin B is used for the prevention or elimination of fungal contaminants in cell culture.

  • Yellow to orange powder
  • Storage temperature of 2 to 8 °C
  • Shelf life of 24 months

Interferes with fungal membrane permeability by forming channels in the membranes and causing small molecules to leak out.

 : This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Synonyms:  (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
  • CAS Number:1397-89-3
  • Formula:C₄₇H₇₃NO₁₇
  • MDL Number:MFCD00877763
  • Molecular Weight:924.08 g/mol

 

Specifications

Specifications

  • Pk:
    250 mg

Specifications