Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, p-Xylene 2,000 µg/ml in methanol
Organic reference standard, p-Xylene 2,000 µg/ml in methanol
Supplier:  Restek
Certificates
Organic reference standard, p-Xylene 2,000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    106-42-3
  • Formula:
    C₈H₁₀
  • Description:
    p-Xylene

Specifications

Frequently Bought Together