Organic reference standard, m-Xylene 200 µg/ml in methanol
Catalog # 123362F
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:108-38-3
- Description:m-Xylene 200 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₈H₁₀
- Description:m-Xylene