Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, m-Xylene 2,000 µg/ml in methanol
 :  Restek
 
Organic reference standard, m-Xylene 2,000 µg/ml in methanol
 :  Restek

 

  • CAS No.:
    108-38-3
  • Formula:
    C₈H₁₀
  • Description:
    m-Xylene

 

Frequently Bought Together