Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, Trichloroethylene 2000 µg/ml in methanol
 :  Restek
 
Organic reference standard, Trichloroethylene 2000 µg/ml in methanol
 :  Restek

 

  • CAS No.:
    79-01-6
  • Formula:
    C₂HCl₃
  • Description:
    Trichloroethylene

 

Frequently Bought Together