Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Methyl ethyl ketone 5000 µg/ml in methanol
Organic reference standard, Methyl ethyl ketone 5000 µg/ml in methanol
Supplier:  Restek
Certificates
Organic reference standard, Methyl ethyl ketone 5000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    78-93-3
  • Formula:
    C₄H₈O 
  • Description:
    Methyl ethyl ketone

Specifications

Frequently Bought Together