Organic reference standard, Chloroform 2000 µg/ml in methanol
: Restek
- CAS No.:67-66-3
- Formula:CHCl₃
- Description:Chloroform
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK