Organic reference standard, 1,1,1-Trichloroethane 100 µg/ml in methanol
Catalog # CMPLC02A.D1.1ML.1
Supplier: CUSTOM MADE CHEMICALS LAB
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1,5 mL
- CAS No.:71-55-6
- Description:1,1,1-Trichloroethane 100 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₂H₃Cl₃
- Description:1,1,1-Trichloroethane