Nickel(II) sulphate hexahydrate, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10101-97-0
:
Formula:NiSO₄·6H₂O
MDL Number:MFCD00149813
:262.85 g/mol
Merck Index:13,06541
Melting Pt:840 °C
Density:2.07 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-104-9
ADR:9,III
:M-Clarity™ quality level = MQ200
UN:3077
:GHS08
:GHS07
:GHS09
Frequently Bought Together