Order Entry
Denmark
ContactUsLinkComponent
Iron(II) sulphate heptahydrate 98.0-105.0% Ph. Eur.
Irritant
Iron(II) sulphate heptahydrate 98.0-105.0% Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Iron(II) sulphate heptahydrate 98.0-105.0% Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    98.0 - 105.0 %
  • Appearance:
    Conforms (see CoA/CoS)
  • Identification (SO₄):
    Passes test
  • Identification (Fe):
    Passes test
  • pH (5 %):
    3.0 - 4.0
  • Cl (Chloride):
    ≤ 200 ppm
  • Elemental impurities (Cr):
    ≤ 50 ppm
  • Elemental impurities (Co):
    ≤ 25 ppm
  • Elemental impurities (Cu):
    ≤ 50 ppm
  • Elemental impurities (Ni):
    ≤ 50 ppm
  • Ferric ions:
    ≤ 0.3 %
  • Mn (Manganese):
    ≤ 0.1 %
  • Zn (Zinc):
    ≤ 50 ppm
  • Residual solvents:
    Unlikely by manuf.process
  • As (Arsenic):
    ≤ 2 ppm
  • Cd (Cadmium):
    ≤ 20 ppm
  • Hg (Mercury):
    ≤ 2 ppm
  • Pb (Lead):
    ≤ 5 ppm
  • V (Vanadium):
    ≤ 2 ppm
  • Conforms to Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together