Iron(III) chloride hexahydrate 99.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10025-77-1
:
Formula:FeCl₃·6H₂O
MDL Number:MFCD00149712
:270.3 g/mol
Merck Index:13,04048
Boiling Pt:280…285 °C (1013 hPa)
Melting Pt:37 °C
Density:1.82 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-729-4
UN:3260
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Assay:99.0 - 102.0 %
- Free chlorine:Passes test
- Heavy metals (as Pb):Max. 50 ppm
- Insoluble substances:Max. 100 ppm
- Total N (Nitrogen):Max. 10 ppm
- Total P (Phosphorus) (as PO4):Max. 100 ppm
- NO3 (Nitrate):Max. 100 ppm
- SO4 (Sulphate):Max. 100 ppm
- Ca (Calcium):Max. 100 ppm
- Cu (Copper):Max. 30 ppm
- Fe (II) (Iron):Max. 20 ppm
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 50 ppm
- Na (Sodium):Max. 0.05 %
- Zn (Zinc):Max. 30 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
Frequently Bought Together