Manganese(II) sulphate monohydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:10034-96-5
:
Formula:MnSO₄·1H₂O
MDL Number:MFCD00149159
Merck Index:13,05763
Melting Pt:700 °C
Density:2.95 g/cm³ (25 °C)
EINECS:232-089-9
UN:3077
ADR:9,III
:169.02 g/mol
:GHS08
:GHS09
Storage Temperature:Refrigerator
Frequently Bought Together