

Manganese(II) sulphate monohydrate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10034-96-5
:
Formula:MnSO₄·1H₂O
MDL Number:MFCD00149159
Merck Index:14,5739
Melting Pt:700 °C
Density:2.95
EINECS:232-089-9
UN:3077
ADR:9,III
:169.02 g/mol
:GHS05
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together