


Cadmium(II) sulphate 8/3-hydrate
Catalog # ROTH4482.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
:
:256.51 g/mol (anhy)
Merck Index:13,01627
Melting Pt:41 °C
Density:3.79 g/cm³ (20 °C)
EINECS:233-331-6
UN:2570
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:7790-84-3
Formula:3CdSO₄·8H₂O
MDL Number:MFCD02253000