Order Entry
Denmark
ContactUsLinkComponent
Bole white (kaolin), TECHNICAL, washed
Bole white (kaolin), TECHNICAL, washed
Catalog # 24926.364
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bole white (kaolin), TECHNICAL, washed
Catalog # 24926.364
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :258.16 g/mol
Merck Index:13,05300
Melting Pt:1760 °C
Density:0.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:310-194-1
CAS Number:1332-58-7
Formula:Al₂SiO₇·2H₂O
MDL Number:MFCD00062311