Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Talc

Glass bottle for solids, 1 kg
Catalog # SWED14450-1
Irritant
Supplier:   SWED HANDLING
Talc
Catalog # SWED14450-1
Supplier:  SWED HANDLING
CAS Number:  14807-96-6
Talc

Glass bottle for solids, 1 kg

CAS Number:  14807-96-6
Price
245,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Magnesium silicate monohydrate, Soapstone, Talcum venetum
  • CAS Number:14807-96-6
  • Formula:Mg₃O₁₁Si₄·1H₂O
  • MDL Number:MFCD00792903
  • Molecular Weight:379.27 g/mol
  • Merck Index:13,09127
  • Melting Pt:>1200 °C
  • Density:2.58…2.83 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:238-877-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications