Order Entry
Denmark
Orders LinkContactUsLinkComponent
Talc, GPR RECTAPUR®

Talc, GPR RECTAPUR®

Plastic bottle for solids, 500 g
Catalog # 28460.268
Supplier:   VWR Chemicals
Talc, GPR RECTAPUR®
Catalog # 28460.268
Supplier:  VWR Chemicals
CAS Number:  14807-96-6
Talc, GPR RECTAPUR®
Talc, GPR RECTAPUR®

Plastic bottle for solids, 500 g

CAS Number:  14807-96-6
Price
195,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Magnesium silicate monohydrate, Soapstone, Talcum venetum
  • CAS Number:14807-96-6
  • Formula:Mg₃O₁₁Si₄·1H₂O
  • MDL Number:MFCD00792903
  • Molecular Weight:379.27 g/mol
  • Merck Index:13,09127
  • Melting Pt:> 1200 °C
  • Density:2.58…2.83 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:238-877-9
Specifications

Specification Test Results

  • Loss on ignition:
    Max. 10 %

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.