Order Entry
Denmark
ContactUsLinkComponent
Calcium chloride dihydrate
Irritant
Calcium chloride dihydrate
Catalog # SERA39551.01
Supplier:  Serva
CAS Number:  
Irritant
Calcium chloride dihydrate
Catalog # SERA39551.01
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :147.01 g/mol
Merck Index:13,01660
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:176 °C
Density:1.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-140-8
 :GHS07
CAS Number:10035-04-8
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613