Order Entry
Denmark
ContactUsLinkComponent
 
Dipotassium hydrogen orthophosphate trihydrate, Millipore®
Catalog # 529567-1
CAS Number:  
Dipotassium hydrogen orthophosphate trihydrate, Millipore®
Catalog # 529567-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :228.22 g/mol
Boiling Pt:∼340 °C (1013 hPa)
Density:2.504 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:16788-57-1
 :7778-77-0
Formula:K₂HPO₄·3H₂O
MDL Number:MFCD00149926