Order Entry
Denmark
ContactUsLinkComponent
Potassium hexacyanoferrate(II) trihydrate ≥98%, GPR RECTAPUR®
Environment
Potassium hexacyanoferrate(II) trihydrate ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Environment
Potassium hexacyanoferrate(II) trihydrate ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 0.02 %

Specification Test Results

Frequently Bought Together