
3,5-Dihydroxytoluene monohydrate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6153-39-5
:
Formula:C₇H₈O₂·1H₂O
MDL Number:MFCD00149092
:142.15 g/mol
Merck Index:14,6864
Boiling Pt:147 °C (5 mmHg)
Melting Pt:56…60 °C
Density:1.29 g/cm³ (4 °C)
Flash Pt:159 °C
EINECS:207-984-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together