Order Entry
Denmark
ContactUsLinkComponent
Sodium acetate trihydrate ≥98%, GPR RECTAPUR®
Sodium acetate trihydrate ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Sodium acetate trihydrate ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together