Hydroxylammonium chloride for synthesis, Sigma-Aldrich®
Catalog # 8.22334.9025
Supplier: Merck
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Grade:Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
- Synonyms:Hydroxylammonium Chloride
- Chemical Formula:NH2OH·HCl
Specification Test Results
About this item
:
:69.49 g/mol
Merck Index:13,04853
Boiling Pt:306 °C (1013 hPa)
Melting Pt:151 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-798-2
UN:2923
ADR:8 (6.1),III
:M-Clarity™ quality level = MQ200
:GHS08
:GHS07
:GHS09
:GHS05
CAS Number:5470-11-1
Formula:NH₂OH·HCl
MDL Number:MFCD00051089