Hydroxylammonium chloride ACS, ISO, Supelco®
Catalog # 1.04619.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay (manganometric):≥ 99.0 %
- Clarity of the solution in Ethanol:passes test
- pH-value (5 %; water):2.5 - 3.5
- Titrable free acid:≤ 0.25 meq/g
- Sulfate (SO₄):≤ 0.002 %
- Heavy metals (ACS):≤ 0.0005 %
- Cu (Copper):≤ 0.001 %
- Fe (Iron):≤ 0.0005 %
- Hg (Mercury):≤ 0.000001 %
- NH₄ (Ammonium):passes test
- Pb (Lead):≤ 0.0005 %
- Total sulfur (as sulfate):≤ 0.005 %
- Residue on ignition (as sulfate):≤ 0.01 %
Specification Test Results
About this item
:
:69.49 g/mol
Merck Index:13,04853
Boiling Pt:306 °C (1013 hPa)
Melting Pt:151 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-798-2
UN:2923
ADR:8,III
:M-Clarity™ quality level = MQ100
:GHS08
:GHS05
:GHS07
:GHS09
CAS Number:5470-11-1
Formula:NH₂OH·HCl
MDL Number:MFCD00051089