Hydroxylammonium chloride ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5470-11-1
:
Formula:NH₂OH·HCl
MDL Number:MFCD00051089
:69.49 g/mol
Merck Index:13,04853
Boiling Pt:306 °C (1013 hPa)
Melting Pt:155…158 °C
Density:1,67 g/cm³ (20 °C)
EINECS:226-798-2
UN:2923
ADR:8,III
:GHS05
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Residue after ignition:≤0.05 %
- Titration with KMnO4:≥96.0 %
- Clarity of alcohol solution:Passes test
- Ammonium (NH4):Passes test
- Iron (Fe):≤5 ppm
- Heavy metals (ICP-OES):≤5 ppm
- Sulfur compounds (as SO4):≤0.005 %
- Titrable free acid:≤0.25 meq/g
Specification Test Results
Frequently Bought Together