Sulphuric acid concentrate for dilution to 1L, 0.05 M H2SO4 (0.1N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Density:1.05 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:3264
ADR:8,III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together