Silver nitrate 5% in aqueous solution
Glass bottle, 250 mL
Catalog # ROTHN053.1
Marketsource
Supplier: Roth Carl
Silver nitrate 5% in aqueous solution
Catalog # ROTHN053.1
Supplier: Roth Carl
Marketsource
CAS Number: 7761-88-8
Jump to:
Product Details & Documents
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- CAS Number:7761-88-8
- Formula:AgNO₃
- MDL Number:MFCD00003414
- Molecular Weight:169.87 g/mol
- Storage Temperature:Ambient
- EINECS:231-853-9
Specifications
Specifications
- Pack type:Glass bottle
- Pk:250 mL



