Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium hydroxide 3% in aqueous solution

Sodium hydroxide 3% in aqueous solution

Ætsende
Leverandør:   Ampliqon A/S
Sodium hydroxide 3% in aqueous solution
Leverandør:  Ampliqon A/S
Certifikater
Sodium hydroxide 3% in aqueous solution
Sodium hydroxide 3% in aqueous solution

3 tilgængelige muligheder nedenfor

Product Details & Documents
Synonymer:  Caustic soda
  • CAS nummer:1310-73-2
  • Formel:NaOH
  • MDL nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Opbevaringstemperatur:Ambient
  • EINECS:215-185-5
  • FN:1824
  • ADR:8,III
Product Family Options

Produktinformation

  • Pack type
    Pk
    Tilgængelighed
    Price
  • Glass bottle
    1 L
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 L
    1 * 1 L - 3.510,00 kr
    3.510,00 kr
    1 * 1 L
       3.510,00 kr

  • Glass bottle
    3 L
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 3 L
    1 * 3 L - 4.310,00 kr
    4.310,00 kr
    1 * 3 L
       4.310,00 kr

  • Glass bottle
    10 L
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 L
    1 * 10 L - 7.100,00 kr
    7.100,00 kr
    1 * 10 L
       7.100,00 kr

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.