Order Entry
Denmark
ContactUsLinkComponent
Silver sulphate 10 g/l in sulphuric acid
Corrosive
Silver sulphate 10 g/l in sulphuric acid
Catalog # ROTHX870.1
Supplier:  Roth Carl
CAS Number:  
undefined
Corrosive
Silver sulphate 10 g/l in sulphuric acid
Catalog # ROTHX870.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

About this item

 :
 :311.8 g/mol
EINECS:233-653-7
UN:1830
ADR:8,II
 :GHS05
CAS Number:10294-26-5
Formula:Ag₂SO₄
MDL Number:MFCD00003407