0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Calcium chloride solutions
Calcium chloride 1 M in aqueous solution
Calcium chloride 1 M in aqueous solution
R040
:
G-Biosciences
:
Calcium chloride 1 M in aqueous solution
R040
:
G-Biosciences
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
147.01 g/mol
:
233-140-8
:
GHS07
:
Ambient
:
10035-04-8
:
CaCl₂·2H₂O
:
MFCD00149613