Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hydrobromic acid 47%

Hydrobromic acid 47%

Glass bottle for solids, 500 g
Catalog # WAKO639-04655
Corrosive
Irritant
Marketsource
Supplier:   Wako
Hydrobromic acid 47%
Catalog # WAKO639-04655
Supplier:  Wako
Marketsource
CAS Number:  10035-10-6
Hydrobromic acid 47%

Glass bottle for solids, 500 g

CAS Number:  10035-10-6
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Hydrogen bromide
  • CAS Number:10035-10-6
  • Formula:HBr
  • MDL Number:MFCD00011323
  • Molecular Weight:80.91 g/mol
  • Density:1.47 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:233-113-0
  • UN:1788
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications