Sulphuric acid 0.01 mol/l (0.02 N) in aqueous solution standardised solution 0.02 N in aqueous solution
Catalog # 35649.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
:
:98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589