

(±)-4-Methyl-2-pentanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-11-2
:
Formula:C₆H₁₄O
MDL Number:MFCD00004550
:102.18 g/mol
Boiling Pt:132 °C (1013 hPa)
Melting Pt:–90 °C
Density:0.8093 g/cm³ (20 °C)
Flash Pt:54 °C
UN:2053
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:203-551-7
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:16.4 g/l (20 °C)
- Melting Point:-60 °C
- Molar Mass:102.18 g/mol
- Boiling Point:130 - 133 °C (1013 hPa)
- Vapor Pressure:7.06 hPa (25 °C)
- Flash Point:41 °C
- Explosion Limit:1 - 5.5 %(V)
- Density:0.81 g/cm3 (20 °C)
- Ignition Point:305 °C
Specification Test Results
Related Information
Frequently Bought Together