(±)-4-Methyl-2-pentanol 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: (±)-Methyl isobutyl carbinol
- CAS Number:108-11-2
- Formula:C₆H₁₄O
- MDL Number:MFCD00004550
- Molecular Weight:102.18 g/mol
- Boiling Pt:132 °C (760 mmHg)
- Melting Pt:–90 °C
- Density:0.8 g/cm³
- Flash Pt:41 °C
- Storage Temperature:Ambient
- EINECS:203-551-7
- UN:2053
- ADR:3,III
5 Options Available Below
Product Details & Documents
Synonyms: (±)-Methyl isobutyl carbinol
- CAS Number:108-11-2
- Formula:C₆H₁₄O
- MDL Number:MFCD00004550
- Molecular Weight:102.18 g/mol
- Boiling Pt:132 °C (760 mmHg)
- Melting Pt:–90 °C
- Density:0.8 g/cm³
- Flash Pt:41 °C
- Storage Temperature:Ambient
- EINECS:203-551-7
- UN:2053
- ADR:3,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4100 to 1.4120 (20°C, 589 nm)
- GC:≥99.0 %
- Specific optical rotation:+28° to +32° (20°C, 589 nm) (c=1 in DMF)
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



