(±)-2-Hexanol
Glass bottle for solids, 1 g
Catalog # ROTH9463.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane
- CAS Number:626-93-7
- Formula:C₆H₁₄O
- MDL Number:MFCD00004585
- Molecular Weight:102.18 g/mol
- Boiling Pt:136 °C (1013 hPa)
- Melting Pt:–23 °C
- Density:0.8287 g/cm³ (0 °C)
- Storage Temperature:Ambient
- EINECS:210-971-4
- UN:2282
- ADR:3,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



