Order Entry
Denmark
Orders LinkContactUsLinkComponent
Paraformaldehyde

Paraformaldehyde

Flammable
Health
Irritant
Marketsource
Supplier:   Roth Carl
Paraformaldehyde
Supplier:  Roth Carl
Marketsource
Certificates
Paraformaldehyde

4 Options Available Below

Product Details & Documents
Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • Merck Index:13,07096
  • Boiling Pt:135…136 °C (13 mmHg)
  • Melting Pt:123 °C
  • Density:1.46 g/cm³ (20 °C)
  • Flash Pt:70 °C
  • Storage Temperature:Ambient
  • UN:2213
  • ADR:4.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg

    Specifications:
    Marketsource
    Marketsource