Paraformaldehyde ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Synonyms: Polyoxymethylene
- CAS Number:30525-89-4
- Formula:(CH₂O)n
- MDL Number:MFCD00133991
- Merck Index:14,7025
- Boiling Pt:135…136 °C (13 mmHg)
- Melting Pt:120…170 °C (dec.)
- Density:1.30
- Flash Pt:71 °C (159 °F)
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2213
- ADR:4.1,III
5 Options Available Below
Product Details & Documents
Synonyms: Polyoxymethylene
- CAS Number:30525-89-4
- Formula:(CH₂O)n
- MDL Number:MFCD00133991
- Merck Index:14,7025
- Boiling Pt:135…136 °C (13 mmHg)
- Melting Pt:120…170 °C (dec.)
- Density:1.30
- Flash Pt:71 °C (159 °F)
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2213
- ADR:4.1,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



