Polyethylene glycol 300, EMPROVE® ESSENTIAL Ph. Eur., SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
Melting Pt:–15…–10 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:220 °C
:300 g/mol
Storage Temperature:Ambient
EINECS:500-038-2
Frequently Bought Together