
Polyethylene glycol 200
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
:200 g/mol
Boiling Pt:? 250 °C (1013 hPa)
Melting Pt:–65 °C
Density:1.125 g/cm³
Flash Pt:171 °C
Storage Temperature:Ambient
EINECS:500-038-2
:GHS07
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4580 to 1.4610 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- Color scale:≤70 APHA
- Hydroxyl value:534 to 591 mg KOH/g
Specification Test Results
Frequently Bought Together