Order Entry
Denmark
ContactUsLinkComponent
Chlorobenzene-[D5] 2000 µg/ml in methanol
Chlorobenzene-[D5] 2000 µg/ml in methanol
Supplier:  Restek
Certificates
Chlorobenzene-[D5] 2000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    3114-55-4
  • Formula:
    C₆D₅Cl
  • Description:
    Chlorobenzene-[D5]

Specifications

Frequently Bought Together