Order Entry
Denmark
ContactUsLinkComponent
 
VWR®, Gelatin
  24350.262
 :  
undefined
VWR®, Gelatin
  24350.262
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • CAS No.:
    9000-70-8
  • Source:
  • Species:
  • Environmentally Preferable:
  • Storage conditions:
    Ambient
  • Form:
    Powder
  • Protein synonyms:
    Gelatine
  • Protein/peptide name:
    Gelatin
  • Grade:
    Technical Grade