Kanadabalsam, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:8007-47-4
:
MDL Number:MFCD00132800
Merck Index:13,00948
Density:0,915 g/cm³ (20 °C)
Flash Pt:62 °C
Storage Temperature:Stue temperatur
EINECS:232-362-2
Frequently Bought Together