Order Entry
Denmark
ContactUsLinkComponent
Native Xanthine oxidase (from Buttermilk)
Native Xanthine oxidase (from Buttermilk)
Certificates
Native Xanthine oxidase (from Buttermilk)

Specifications

  • CAS No.:
    9002-17-9
  • Enzyme type:
    Native
  • Source:
    Buttermilk
  • Enzyme name:
    Xanthine oxidase

Specifications

Frequently Bought Together