Platinum rhodium alloy (90:10 w%) ≥99.9% (metals basis), gauze woven from wire 80 mesh, Ø 0.076 mm (0.003 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00198186
Storage Temperature:Ambient
Related Information
Frequently Bought Together