Starch (from rice)
Marketsource
Supplier: Roth Carl
Jump to:
About this item
- CAS Number:9005-25-8
- Formula:(C₆H₁₀O₅)n
- MDL Number:MFCD00082026
- Molecular Weight:162.14 g/mol
- Storage Temperature:Ambient
- EINECS:232-679-6
2 Options Available Below
Product Details & Documents
- CAS Number:9005-25-8
- Formula:(C₆H₁₀O₅)n
- MDL Number:MFCD00082026
- Molecular Weight:162.14 g/mol
- Storage Temperature:Ambient
- EINECS:232-679-6
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



